Learn More
Electron Microscopy Sciences Rotary Diamond Micro-Engraver Pen
Supplier: Electron Microscopy Sciences 5046515
Diamond Micro-Engraver Pen can be used to mark samples, engrave tools, indicate a sample area for finding in the SEM, deburring and so much more. Easy to handle and control, it will engrave --wood, metal, ceramic, glass and more. The micro engraver features a contoured grip for comfort and control and a precision-ground stainless steel bit with a diamond point tip. This diamond-tipped ballpoint micro engraver is also as small as a pen, so it easily fits in your pocket or purse for easy transport. Requires two AAA batteries, included
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.