Learn More
BUCHI Rotavapor™ R-300 Rotary Evaporator with Controller
SDP
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor™ R-300 to a fully integrated system where a central interface regulates each component.
Supplier:  BUCHI 11R300152C122
 View more versions of this product
Catalog No. 05-000-463
Description
Description
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Specifications
Specifications
Manual Lift
5 L
Safety Coating
220°C
100/120 V
607 x 429 x 947 mm
1100 W
Rotary Evaporator
R-300
100/120 V
10 to 280 rpm
B-305 Heating Bath
1600 W
Cold Trap
I-300 Interface (Woulff Bottle)
CE/CSA
50/60 Hz
24/40
Product Suggestions
Product Suggestions