Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotenone 97%, Thermo Scientific™

Catalog No. p-191147
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 83-79-4
Molecular Formula C23H22O6
Molecular Weight (g/mol) 394.423
InChI Key JUVIOZPCNVVQFO-HBGVWJBISA-N
Synonym rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
PubChem CID 6758
ChEBI CHEBI:28201
SMILES CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC

Specifications

Melting Point 156°C to 163°C
Color Yellow to Beige
Infrared Spectrum Authentic
Quantity 5 g
Beilstein 19, II, 438
Merck Index 15, 8403
Formula Weight 394.41
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Rotenone, 97%
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-501-9
RTECSNumber DJ2800000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.