Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotigotine hydrochloride, Tocris Bioscience™

Catalog No. p-4816647 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dopamine D2/D3 agonist

Dopamine D2 and D3 receptor agonist. Ki values are 13 and 0.71 nM for D2 and D3 respectively. Also has significant affinity for 5-HT1A and adrenergic α2B receptors. Exhibits antiparkinsonian acitivity.

Chemical Identifiers

CAS 125572-93-2
Molecular Formula C19H26ClNOS
Molecular Weight (g/mol) 351.933
InChI Key CEXBONHIOKGWNU-NTISSMGPSA-N
Synonym rotigotine hydrochloride, rotigotine hcl, s-6-propyl 2-thiophen-2-yl ethyl amino-5,6,7,8-tetrahydronaphthalen-1-ol hydrochloride, 6s-6-propyl 2-thiophen-2-ylethyl amino-5,6,7,8-tetrahydronaphthalen-1-ol hydrochloride, rotigitine, n-0923 hydrochloride, neupro hydrochloride salt, neupro hydrochloride
PubChem CID 180335
IUPAC Name (6S)-6-[propyl(2-thiophen-2-ylethyl)amino]-5,6,7,8-tetrahydronaphthalen-1-ol;hydrochloride
SMILES CCCN(CCC1=CC=CS1)C2CCC3=C(C2)C=CC=C3O.Cl

Specifications

Quantity 10 mg
Formula Weight 351.93
Percent Purity >99%
Chemical Name or Material Rotigotine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.