Learn More
Research Products International Corp Roto-Rock Genie 120V
SDP
Supplier:  Research Products International Corp 150620
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Programable all-in-one benchtop GENIE Rocker / Rotator or cycle from Rock to Rotate. Includes Magnetic Platform and Non-slip Rocking Tray. GENIE clip plate systems designed to handle a variety of vessels from PCR tube, vials, bottels, and sealed bags. Digital set/control for RPM, Total Time, Cycle Time. Feature Pause/Resume button allowing the operator to add reagent to a sample during process.
Catalog No. 50-269-4215
missing translation for 'QuantityLeft'