Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rottlerin, Tocris Bioscience™

Catalog No. p-4785451 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reported PKCδ inhibitor

Originally reported to inhibit PKC isoforms. Also reported to inhibit CAM kinase III. However, recently shown to inhibit a wide range of protein kinases, and most potently to inhibit PRAK and MAPKAP-K2 (IC50 values are 1.9 and 5 μM respectively). Also shown to act as a direct mitochondrial uncoupler. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways.

Chemical Identifiers

CAS 82-08-6
Molecular Formula C30H28O8
Molecular Weight (g/mol) 516.546
InChI Key DEZFNHCVIZBHBI-ZHACJKMWSA-N
Synonym rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
PubChem CID 5281847
ChEBI CHEBI:8899
IUPAC Name (E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one
SMILES CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O

Specifications

Quantity 10 mg
Formula Weight 516.55
Percent Purity >98%
Chemical Name or Material Rottlerin
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.