Learn More
TARGETMOL CHEMICALS INC ROTUNDINE 500MG
Supplier:  TARGETMOL CHEMICALS INC T6648
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rotundine (Gindarine) (L-tetrahydropalmatine L-THP) is a selective dopamine D1 receptor antagonist with IC50 of 166 nM. purity: 99%
Catalog No. 50-257-9319
missing translation for 'QuantityLeft'