Learn More
Medchemexpress LLC HY-N0096 50mg Medchemexpress, Rotundine CAS:483-14-7 Purity:>98%
Supplier: Medchemexpress LLC HYN009650MG
Medchemexpress, HY-N0096 50mg Rotundine CAS:483-14-7 Rotundine is an antagonist of dopamine D1 , D2 and D3 receptors with IC 50 s of 166 nM, 1.4 μM and 3.3 μM, respectively. Rotundine is also an antagonist of 5-HT 1A with an IC 50 of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.