Learn More
Enzo Life Sciences Adenosine-3',5'-cyclic Monophosphothioate, Rp-Isomer . sodium salt (1mg). CAS: 73208-40-9
Supplier: Enzo Life Sciences ALX480085M001
Competitive inhibitor of cyclic AMP-dependent protein kinase I and II. Metabolic stability towards mammalian cyclic nucleotide- responsive phosphodiesterases. Discriminates between protein kinase A (antagonist) and some other cyclic AMP receptors, e.g. channels or CAP 3 (agonist). Membrane-permeant for several systems (for improved permeability more lipophilic analogues e.g. Rp-8-Br-cAMPS (BML-CN216) are recommended). Alternative name: Rp-cAMPS . Na, cAMPS . Na, Rp-Isomer. Purity: ≥99% (HPLC) (low adenosine, cAMP and Sp-cAMPS content). Appearance: White to off-white powder. Please keep in mind that equal amounts of the compound may look different in volume depending on humidity. Rp-cAMPS is hygroscopic and tends to form a transparent film on the bottom of the tube. Solubility: Soluble in water or aqueous buffers, DMSO, Methanol. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.