Learn More
United States Biological Corporation RPMI 1640MED W/O AM 10L
SDP
Supplier:  United States Biological Corporation R899904A10L
Encompass_Preferred
RPMI-1640 was developed by Moore, et al., at Roswell Park Memorial Institute, hence the acronym RPMI.�The formulation is based on the RPMI-1630 series of media utilizing a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.�RPMI-1640 medium has been used for the culture of human normal and neoplastic leukocytes.�RPMI-1640, when properly supplemented, has demonstrated wide applicability for supporting growth of many types of cultured cells, including fresh human lymphocytes in the 72 hour phytohemaglutinin (PHA) stimulation assay.
Catalog No. NC0750940
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.