Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 102895 hydrochloride, Tocris Bioscience™

Catalog No. p-4784448 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CCR2b chemokine receptor antagonist

CCR2-selective chemokine receptor antagonist (IC50 values are 0.36 and 17.8 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Blocks MCP-1-stimulated calcium influx and chemotaxis with IC50 values of 32 nM and 1.7 μM respectively. Also inhibits α1A, α1D and 5-HT1A receptors.

Chemical Identifiers

CAS 1173022-16-6
Molecular Formula C21H22ClF3N2O2
Molecular Weight (g/mol) 426.864
InChI Key KRRISOFSWVKYBF-UHFFFAOYSA-N
PubChem CID 16759153
IUPAC Name 1'-[2-[4-(trifluoromethyl)phenyl]ethyl]spiro[1H-3,1-benzoxazine-4,4'-piperidine]-2-one;hydrochloride
SMILES C1CN(CCC12C3=CC=CC=C3NC(=O)O2)CCC4=CC=C(C=C4)C(F)(F)F.Cl

Specifications

Quantity 10 mg
Synonym rs 102895 hydrochloride, rs102895 hydrochloride, rs-102895 hydrochloride, 1'-4-trifluoromethyl phenethyl spiro benzo d 1,3 oxazine-4,4'-piperidin-2 1h-one hydrochloride, 1'-2-4-trifluoromethyl phenyl ethyl-spiro 4h-3,1-benzoxazine-4,4'-piperidin-2 1h-one hydrochloride, c21h21f3n2o2.clh, rs 102895 hydrochloride hplc , powder, 1 inverted exclamation marka-2-4-trifluoromethyl phenyl ethyl-spiro 4h-3,1-benzoxazine-4,4 inverted exclamation marka-peperidin-2 1h-one, 1'-2-4-trifluoromethyl phenyl ethyl spiro 1h-3,1-benzoxazine-4,4'-piperidine-2-one hydrochloride
Formula Weight 426.86
Percent Purity >99%
Chemical Name or Material RS 102895 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.