Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RS 17053 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
α1A-adrenoceptor antagonist, with very high affinity for α1A receptors (pKi and pA2 estimates of 9.1 - 9.9) and a 30 - 100 fold selectivity over the α1B and α1D subtypes (pKi and pA2 estimates 7.7 - 7.8).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 449.42 |
Percent Purity | >98% |
Chemical Name or Material | RS 17053 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction