Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 79948 hydrochloride, Tocris Bioscience™

Catalog No. p-4785479 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective α2 antagonist

α2-adrenoreceptor antagonist (Kd values are; 0.18, 0.19 and 0.42 nM for α2B α2C and α2A, respectively in rat).

Chemical Identifiers

CAS 186002-54-0
Molecular Formula C19H29ClN2O3S
Molecular Weight (g/mol) 400.962
InChI Key DZTZUOBWDBPPJQ-BQBHMPFISA-N
Synonym rs 79948 hydrochloride, 8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride, 4ar,12bs,13as-1-ethanesulfonyl-10-methoxy-2,3,4,4a,5,7,8,12b,13,13a-decahydro-1,6-diazatetraphene hydrochloride, 8ar,12as,13as-12-ethylsulfonyl-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride, 8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride
PubChem CID 9908991
IUPAC Name (8aR,12aS,13aS)-12-ethylsulfonyl-3-methoxy-5,6,8,8a,9,10,11,12a,13,13a-decahydroisoquinolino[2,1-g][1,6]naphthyridine;hydrochloride
SMILES CCS(=O)(=O)N1CCCC2C1CC3C4=C(CCN3C2)C=C(C=C4)OC.Cl

Specifications

Quantity 10 mg
Formula Weight 400.96
Percent Purity >99%
Chemical Name or Material RS 79948 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.