Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rufinamide, Tocris Bioscience™

Catalog No. p-4816648 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Prolongs inactivation of sodium channels; anticonvulsant

Board spectrum anticonvulsant. Prolongs the inactivation of sodium channels and limits the frequency of action potential firing in cultured and acutely isolated neurons. Displays anticonvulsive activity in a range of animal seizure models.

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O
Molecular Weight (g/mol) 238.198
InChI Key POGQSBRIGCQNEG-UHFFFAOYSA-N
Synonym rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
PubChem CID 129228
IUPAC Name 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMILES C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F

Specifications

Quantity 10 mg
Formula Weight 238.19
Percent Purity >99%
Chemical Name or Material Rufinamide
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.