Learn More
TARGETMOL CHEMICALS INC RUNCACIGUAT 5MG
Supplier:  TARGETMOL CHEMICALS INC T16808
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Runcaciguat is a soluble guanylate cyclase stimulator. Runcaciguat combines with selective partial adenosine A1 receptor agonists and can be used in studies about cardiovascular and renal diseases. purity: 98%
Catalog No. 50-258-1149
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.