Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RX 821002 hydrochloride, Tocris Bioscience™

Catalog No. p-4785487 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective α2D antagonist

Potent, selective α2-adrenoceptor antagonist with very low affinity for imidazoline sites. Displays selectivity for the α2D over the α2A subtypes (pKd values are 9.7 and 8.2 respectively).

Chemical Identifiers

CAS 109544-45-8
Molecular Formula C12H15ClN2O3
Molecular Weight (g/mol) 270.713
InChI Key IMPOOMVZVWKSAP-UHFFFAOYSA-N
Synonym rx 821002 hydrochloride, 2-2,3-dihydro-2-methoxy-1,4-benzodioxin-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-2-2-methoxy-1,4-benzodioxanyl imidazoline hydrochloride, 2-3-methoxy-2h-1,4-benzodioxin-3-yl-4,5-dihydro-1h-imidazole,hydrochloride, 2-2-methoxy-3h-1,4-benzodioxin-2-yl-4,5-dihydro-1h-imidazole hydrochloride, rx 821002 hydrochloride, solid hplc, 2-2-2-methoxy-1,4-benzodioanyl-imidazoline hydrochloride, hydrochloride 2-2-methoxy-1,4-benzodioxan-2-yl-2-imidazoline, 2-2-methoxy-2,3-dihydrobenzo b 1,4 dioxin-2-yl-4,5-dihydro-1h-imidazole hydrochloride
PubChem CID 11957683
IUPAC Name 2-(3-methoxy-2H-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES COC1(COC2=CC=CC=C2O1)C3=NCCN3.Cl

Specifications

Quantity 10 mg
Formula Weight 270.72
Percent Purity >99%
Chemical Name or Material RX 821002 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.