Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline 98.0+%, TCI America™
SDP

Catalog No. p-7123352 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 117408-98-7
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD22200533
InChI Key JDOAIDZRVWJBEG-SNVBAGLBSA-N
Synonym (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole
PubChem CID 14337774
IUPAC Name (4S)-4-tert-butyl-2-pyridin-2-yl-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C)C1COC(=N1)C2=CC=CC=N2

Specifications

Melting Point 72°C
Color White
Quantity 1 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-tert-Butyl-2-(2-pyridyl)oxazoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.