Learn More
Bellco Glass, Inc. Nephelo Flask 3 Bfl Sc 500ML 19X130Mm
SDP
Supplier:  Bellco Glass, Inc. 258119135
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Has a threaded neck and sidearm with clean-out port. 3 baffles increase oxygenation of cultures. Supplied with 38mm and 18mm rubb
Catalog No. 50-194-5586
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.