Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-(+)-Dimethindene maleate, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
(S)-(+)-Dimethindene maleate is an enantiomer that is a subtype-selective M2 muscarinic receptor antagonist (pKi values are 7.08, 7.78, 6.70 and 7.00 for M1, M2, M3 and M4 receptors respectively). Also H1 histamine receptor antagonist (pKi = 7.48). Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), minocycline hydrochloride (Cat.No. 3268) and human leukemia inhibitory factor.
For more information about how Dimethindene maleate may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 408.5 |
Percent Purity | >99% |
Chemical Name or Material | (S)-(+)-Dimethindene maleate |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction