Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-Glycidylphthalimide 98.0+%, TCI America™
SDP

Catalog No. p-7130241 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 161596-47-0
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD04973350
InChI Key DUILGEYLVHGSEE-ZETCQYMHSA-N
Synonym (S)-N-(2,3-Epoxypropyl)phthalimide
PubChem CID 719412
IUPAC Name 2-[[(2S)-oxiran-2-yl]methyl]isoindole-1,3-dione
SMILES C1C(O1)CN2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 203.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-Glycidylphthalimide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.