Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-p-Tolyl p-Toluenesulfonothioate 98.0+%, TCI America™
SDP

Catalog No. p-7137620 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 2943-42-2
Molecular Formula C14H14O2S2
Molecular Weight (g/mol) 278.38
MDL Number MFCD00092126
InChI Key HSAROCRPZOYKGS-UHFFFAOYSA-N
Synonym p-Toluenesulfonothioic Acid S-p-Tolyl Ester
PubChem CID 257647
IUPAC Name 1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene
SMILES CC1=CC=C(SS(=O)(=O)C2=CC=C(C)C=C2)C=C1

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Formula Weight 278.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material S-p-Tolyl p-Toluenesulfonothioate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.