Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. p-7119560 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
500 mg
5 g

Chemical Identifiers

CAS 118949-61-4
Molecular Formula C17H23N3O2
Molecular Weight (g/mol) 301.39
MDL Number MFCD00191815
InChI Key CSGQGLBCAHGJDR-HUUCEWRRSA-N
Synonym (S,S)-2,2′C-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)
PubChem CID 688211
IUPAC Name (4S)-4-propan-2-yl-2-[6-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)C1COC(=N1)C2=NC(=CC=C2)C3=NC(CO3)C(C)C

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 500 mg
Formula Weight 301.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.