Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-Sulpiride, Tocris Bioscience™

Catalog No. 0895100 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard selective D2-like antagonist

(S)-(-)-Sulpiride is an active enantiomer. Selective D2-like dopamine antagonist (Ki values are ∼ 0.015, ∼ 0.013, 1, ∼ 45 and ∼ 77 μM at D2, D3, D4, D1 and D5 receptors respectively).

Racemate also available.

Chemical Identifiers

CAS

23672-07-3

Molecular Formula

C15H23N3O4S

Molecular Weight (g/mol)

341.426

InChI Key

BGRJTUBHPOOWDU-NSHDSACASA-N

Synonym

levosulpiride, --sulpiride, s---sulpiride, levopraid, levopride, l-sulpiride, unii-jtg7r315lk, s-sulpiride, levosulpirida

PubChem CID

688272

ChEBI

CHEBI:64119

IUPAC Name

N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-methoxy-5-sulfamoylbenzamide

SMILES

CCN1CCCC1CNC(=O)C2=C(C=CC(=C2)S(=O)(=O)N)OC

Specifications

Quantity

100 mg

Formula Weight

341.42

Percent Purity

>99%

Chemical Name or Material

(S)-(-)-Sulpiride

Recommended Storage

Store at Room Temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.