Learn More
Cayman Chemical Sphingosine-1-phosphate (d18:1), 26993-30-6, 1 mg
SDP
Supplier:  Cayman Chemical 625701
Encompass_Preferred
Molecular Formula C18H38NO5P, Purity ≥98%, Formula Weight 379.5, Synonyms: S1P (d18:1), Sphingosine-1-Phosphoric Acid; A potent lipid signaling molecule that exhibits a wide range of biological activities; it enhances cell growth and inhibits the normal apoptotic response to a variety of stimuli.
Catalog No. NC9799531
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.