Learn More
NEOGEN Sabouraud Dextrose Agar, Emmons, 500 g
SDP
Supplier:  NEOGEN 700003620
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Sabouraud Dextrose Agar, Emmons is used for the cultivation of fungi in a laboratory setting. Sabouraud Dextrose Agar, Emmons is not intended for use in the diagnosis of disease or other conditions in humans. Sabouraud Dextrose Agar (SDA), Emmons is a modification of Dextrose Agar described by Sabouraud. SDA is used for cultivating pathogenic and commensal fungi and yeasts. Emmons modified the formula by adjusting the pH and reducing the dextrose concentration. This neutral pH enhances recovery of some pathogenic fungi. Sabouraud Dextrose Agar, Emmons is an excellent basal medium, and antibiotics and inhibitors may be added for the selective cultivation and isolation of various microorganisms.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-agar-emmons
Catalog No. 50-201-5906