Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Sacubitril calcium
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Sacubitril calcium is a membrane metallo-endopeptidase (neprilysin) inhibitor prodrug. Increases ANF-induced natriuresis without affecting diuresis in rats. Orally bioavailable.
Specifications
Specifications
Chemical Name or Material | Ethyl (αR,γS)-γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-[1,1′-biphenyl]-4-pentanoate hemicalcium salt |
CAS | 1369773-39-6 |
Synonym | AHU 377 |
Quantity | 10 mg |
Target | Additional Synthetic Protease Inhibitors |
Molecular Formula | C24H28NO5.1/2Ca |
Formula Weight | 430.52 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction