Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Safe-Sense™ Bouffant Caps
SDP

Catalog No. p-7224311
Click to view available options
Style:
Pleated
Traditional

Bouffants are manufactured from a strong, lightweight, and breathable spun-bounded polypropylene (SPP) fabric to protect against dirt, grime, and certain dry particulates in controlled environments.

  • Bouffant caps
  • Used for cleanroom
  • Non-sterile
  • 21 in. diameter
  • Choose from pleated or traditional style

Specifications

Product Type Cleanroom Bouffant
Size 21 in.
Material Spunbond Polypropylene
Color White
Disposable Single-use
Model 5971-01
Sterility Non-sterile
Certifications/Compliance Manufactured in an ISO 13485 facility
For Use With (Application) Controlled Environments
Style Pleated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.