Learn More
Pyramex™ Safety Choice Perforated Goggles
Perforated Safety Choice Perforated Goggles provide excellent ventilation. Lenses provide 99.9% protection against harmful UV rays.
$28.70
Specifications
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Perforated Goggles
Material Polycarbonate
Disposable Reusable
View More Specs
Products 1
Catalog Number Mfr. No. Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Price Quantity & Availability  
19-817-306
View Documents
Pyramex™
G201
Scratch-resistant, Impact-resistant, UV-treated
Case of 12 for $28.70
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Perforated
  • Manufactured from clear polycarbonate
  • Scratch resistant
  • 99% UVA/B absorption
Warranty and Services
90 Days
Specifications
Specifications
ANSI Z87.1
Perforated Goggles
Reusable
PVC
Wrap-around
Clear
Polycarbonate
Polycarbonate
U6
Not Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.