Learn More
TARGETMOL CHEMICALS INC SAFINAMIDE 5MG
Supplier:  TARGETMOL CHEMICALS INC T6651L
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Safinamide (FCE-26743) is a reversible and selective monoamine oxidase B (IC50 0.098uM) inhibitor reducing the degradation of dopamine and a glutamate release inhibitor (IC50 8uM). It also inhibits dopamine reuptake. Safinamide also blocks sodium and calcium channels. Potential additional uses of Safinamide might be Parkinson's disease restless leg syndrome and epilepsy. purity: 99%
Catalog No. 50-257-9317
missing translation for 'QuantityLeft'