Learn More
Enzo Life Sciences SAG, (1mg), CAS Number: 364590-63-6
SDP
Supplier:  Enzo Life Sciences ALX270426M001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Smoothened ligand. A potent cell-permeable benzothiophene compound that modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM). Shown to induce Hedgehog pathway activation (EC50= ~3 nM in NIH 3T3-derived Shh-LIGHT2 cells) and counteracts Cyclopamine (BML-GR334) inhibition of Smo. Reported to act as an activator at low concentrations and as an inhibitor at very high concentrations. Formula: C28H28ClN3OS. MW: 490.1. CAS Number: 364590-63-6. Purity: ≥95% (HR-MS, 1H-NMR, 13C-NMR). Appearance: White to off-white solid. Solubility: Soluble in DMSO (10mg/ml), sparingly soluble in water (acidification with HCl increases water solubility). Use/Stability: Stable for at least 6 months when stored at -20°C. Long Term Storage: -20°C
Catalog No. NC9953751
missing translation for 'QuantityLeft'