Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Salbutamol hemisulfate, 98%

Catalog No. AC456460050
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C13H21NO3·1/2H2SO4
Molecular Weight (g/mol) 288.35
InChI Key OVICLFZZVQVVFT-UHFFFAOYSA-N
Synonym salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
PubChem CID 9884233
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
SMILES CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O

Specifications

Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 288.35
Percent Purity 98%
Chemical Name or Material Salbutamol hemisulfate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 256-916-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.