Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid, For ACS analysis, 99.68%, MP Biomedicals™

Catalog No. p-4404949 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
500 g

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Color White
Assay Percent Range >99%
Formula Weight 138.12
Density 1.443g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Salicylic acid, For ACS analysis
EINECSNumber 200-712-3
Recommended Storage Store at room temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.