Learn More
Rockland Immunochemicals Salmon Sperm DNA Ready-to-Use Solution (Packaged 5 x 5 mg) - MB-103-0025
SDP
Supplier:  Rockland Immunochemicals MB1030025
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Prepared by a modification of the method of Emanuel and Chaikoff, JBC 203, 164 (1953) from salmon testes DNA by mechanical shearing and heat denaturation to an average size of 100 to 200 base pairs. To reverse any renaturation occurring during storage, this material should be briefly boiled and rapidly chilled prior to use. >75% native nucleic acid.
Catalog No. NC0248895
missing translation for 'QuantityLeft'