Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7049527
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13465-58-2
MDL Number MFCD00011236

Specifications

Melting Point 450°C (in -8H2O)
Physical Form Crystalline
Quantity 25 g
Merck Index 14,8347
Solubility Information Sparingly soluble in water
Formula Weight 733.00 (588.88 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Samarium(III) sulfate octahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-211-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.