Learn More
Bioworld SAMBUCUS NIGRA LECTIN - FITC
Supplier:  Bioworld 217610511
Encompass_Preferred
Specification: Sambucus nigra lectin (SNA-I) from elderberry, FITC conjugated.; Carbohydrate specificity: NANA(Neu5Ac(2,6)Gal/GalNAc; Inhibitory carbohydrate: High concentration of lactose, -Galactose; Extremely light sensitive (fluoresc...
Catalog No. 50-199-160
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.