Learn More
Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
Supplier: Thermo Scientific™ 03 0181
Description
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Available Options:
- Nonsterile or gamma irradiation sterilized
- Labled or non-labled
- Bulk or individually packed
Specifications
60 mL | |
Yes | |
Specimen Container | |
60mL | |
Non-sterile | |
Cap and container bagged separately | |
w/o Label | |
500 |
48 mm | |
Cylinder | |
Virgin Polypropylene | |
White | |
Bio-Tite | |
Cap and container bagged separately, non-sterile, no label, white caps | |
48mm | |
500 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.