Learn More
Anton Paar Kit to help prepare samples for measurment in the Alex 500
Supplier:  Anton Paar 165008
Encompass_Preferred
Alex 500 sample preparation kit contains contains 2 Erlenmeyer flasks with rubber stoppers, 1 beaker, 1 funnel, 2 spoons, 100 folded paper filters, 1 kg of diatomaceous earth and 1 dust mask.Recommended for use with Anton Paar Alex 500 Alcohol and Extract Meter for Beer, Wine, Cider, Spirits, Liqueurs, and Sake NC1818206
Catalog No. NC1818625
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.