Learn More
Electron Microscopy Sciences Flexible Shaft Machine Sanding Discs, Coarse Grit, 3/4" diameter
Supplier:  Electron Microscopy Sciences 6214510
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Coarse
Diameter: 3/4"
Catalog No. 50-996-832
missing translation for 'QuantityLeft'