Learn More
Electron Microscopy Sciences Flexible Shaft Machine Sanding Discs, Fine Grit, 7/8" diameter
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Fine
Diameter: 7/8"