Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Santonin, MP Biomedicals™

Catalog No. p-7083125 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10g
50g

Santonin, MP Biomedicals is a chiral building block used for the synthesis of various sesquiterpenoides.

Chemical Identifiers

CAS 481-06-1
Molecular Formula C15H18O3
Molecular Weight (g/mol) 246.31
MDL Number MFCD00135865
InChI Key XJHDMGJURBVLLE-BOCCBSBMSA-N
Synonym santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum
PubChem CID 221071
ChEBI CHEBI:16363
IUPAC Name (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
SMILES C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O

Specifications

Melting Point 172°C to 173°C (literature)
Quantity 10g
Solubility Information Soluble in cold water, boiling water, alcohol cold/boiling, cold/boling ether, chloroform (50mg/mL).
Formula Weight 246.31
Optical Rotation [α]20/D −170 ± 5° (c=1% in ethanol) (Lit.)
Physical Form Off-White Crystalline Powder
Percent Purity ≥98%
Recommended Storage 4°C, desiccate, protect from light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.