Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 258719 hydrochloride, Tocris Bioscience™

Catalog No. p-4784490 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-HT7 antagonist

Selective 5-HT7 receptor antagonist that displays > 100-fold selectivity over a range of other receptors. Reverses the hypothermic effect of 5-CT in mice following i.p. administration.

Chemical Identifiers

CAS 1217674-10-6
Molecular Formula C18H31ClN2O2S
Molecular Weight (g/mol) 374.968
InChI Key UIZKHTBWJSUGOV-UNTBIKODSA-N
Synonym sb 258719 hydrochloride, 3-methyl-n-1r-1-methyl-3-4-methyl-1-piperidinyl propyl-n-methylbenzenesulfonamide hydrochloride, n,3-dimethyl-n-2r-4-4-methylpiperidin-1-yl butan-2-yl benzenesulfonamide hydrochloride
PubChem CID 56972181
IUPAC Name N,3-dimethyl-N-[(2R)-4-(4-methylpiperidin-1-yl)butan-2-yl]benzenesulfonamide;hydrochloride
SMILES CC1CCN(CC1)CCC(C)N(C)S(=O)(=O)C2=CC=CC(=C2)C.Cl

Specifications

Quantity 10 mg
Formula Weight 374.97
Percent Purity >99%
Chemical Name or Material SB 258719 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.