Learn More
Medchemexpress LLC HY-50688 25mg Medchemexpress, SB-265610 CAS:211096-49-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5068825MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-50688 25mg SB-265610 CAS:211096-49-0 SB-265610 is a selective, competitive, nonpeptide and allosteric CXCR2 antagonist. SB-265610 blocks rat cytokine-induced neutrophil chemoattractant-1 (CINC-1)-induced calcium mobilization and neutrophil chemotaxis with IC50s of 3.7 nM and 70 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0071
missing translation for 'QuantityLeft'