Learn More
TARGETMOL CHEMICALS INC SB-277011 10MG
Supplier:  TARGETMOL CHEMICALS INC T12861
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-277011 (SB-277011A) is a specific antagonist of the dopamine D3 receptor with pKi values of 8.0 6.0 5.0 and <5.2 for D3 receptor D2 receptor 5-HT1D and 5-HT1B respectively. purity: 98%
Catalog No. 50-258-1409
missing translation for 'QuantityLeft'