Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 277011A dihydrochloride, Tocris Bioscience™

Catalog No. p-4816903 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D3 antagonist

Selective dopamine D3 receptor antagonist (pKi values are 8.0, 6.0, 5.0 and <5.2 for D3, D2, 5-HT1D and 5-HT1B respectively). Brain penetrant.

Chemical Identifiers

CAS 1226917-67-4
Molecular Formula C28H32Cl2N4O
Molecular Weight (g/mol) 511.491
InChI Key HEZIOTGUXSPDAK-UHFFFAOYSA-N
Synonym sb 277011a dihydrochloride, c28h30n4o.2hcl, n-1r,4r-4-2-6-cyano-3,4-dihydroisoquinolin-2 1h-yl ethyl cyclohexyl quinoline-4-carboxamide dihydrochloride, n-trans-4-2-6-cyano-3,4-dihydro-2 1h-isoquinolinyl ethyl cyclohexyl-4-quinolinecarboxamide dihydrochloride, n-4-2-6-cyano-3,4-dihydro-1h-isoquinolin-2-yl ethyl cyclohexyl quinoline-4-carboxamide dihydrochloride
PubChem CID 75358288
IUPAC Name N-[4-[2-(6-cyano-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]cyclohexyl]quinoline-4-carboxamide;dihydrochloride
SMILES C1CC(CCC1CCN2CCC3=C(C2)C=CC(=C3)C#N)NC(=O)C4=CC=NC5=CC=CC=C45.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 511.49
Percent Purity >98%
Chemical Name or Material SB 277011A dihydrochloride
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.