Learn More
As One International Inc 10x Standard Buffer, Triton free, 10 x 1.5 ml
Supplier:  As One International Inc AO302410
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry.
Catalog No. 50-165-5973
missing translation for 'QuantityLeft'