Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SC 236, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective COX-2 inhibitor (IC50 values are 0.005 and 17.8 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory properties and potent antimetastatic activity against both experimental metastases and spontaneous metastases arising following primary tumor excision.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 401.79 |
Percent Purity | >99% |
Chemical Name or Material | SC 236 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction