Learn More
Sklar Instruments Scalpel Handle, Sklar 5-3/4 in., Straight, Round, #3R / Knurled, Stainless Steel. EA.
SDP
Supplier:  Sklar Instruments 062908
Encompass_Preferred
Scalpel Handle, Sklar. 5-3/4 in., Straight, Round, #3R / Knurled, Stainless Steel. Sklar's #3R Round Scalpel Handle is a straight handle that can be used with scalpel blades #10, #11, #12, and #15.
Catalog No. 50-118-1200
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.