Learn More
Scandium(III) acetate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Specifications
Linear Formula | (CH3CO2)3Sc·xH2O |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 222.10 (Anhydrous) |
Percent Purity | 99.9% |
Assay | (metals basis) |
Chemical Name or Material | Scandium(III) acetate hydrate |
Safety and Handling
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.