Learn More
SCF/c-kit Ligand Rabbit anti-Human, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | SCF/c-kit Ligand |
---|---|
Clone | 4 |
Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
SCF/c-kit Ligand Monoclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
SCF/c-kit Ligand | |
ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
Recombinant Human SCF/c-kit Ligand protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
4 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
4254 | |
IgG | |
Protein A purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.