Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCH 23390 hydrochloride, Tocris Bioscience™

Catalog No. p-4785549 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard selective D1-like antagonist; also 5-HT2C agonist and Kir3 channel blocker

Potent dopamine receptor antagonist (Ki values are 0.2 nM and 0.3 nM at D1 and D5 receptor sub-types, respectively). Also an agonist at 5-HT2C receptors in vitro (Ki values are 6.3 - 9.3 nM). Blocks quinpirole-induced Kir3 (GIRK) currents (EC50 = 268 nM) independently of receptors.

Chemical Identifiers

CAS 125941-87-9
Molecular Formula C17H19Cl2NO
Molecular Weight (g/mol) 324.245
InChI Key OYCAEWMSOPMASE-XFULWGLBSA-N
Synonym r +-sch-23390 hydrochloride, sch 23390 hydrochloride, sch-23390 hydrochloride, unii-4s8t3e2f4z, r-+-sch 23390 hydrochloride, 5r-8-chloro-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrochloride, r +-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, r-+-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, sch23390 hydrochloride
PubChem CID 11957535
IUPAC Name (5R)-8-chloro-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrochloride
SMILES CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 324.24
Percent Purity >98%
Chemical Name or Material SCH 23390 hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.